Cole-Parmer® Muffle Furnaces with MoSi2 Heating Elements
Muffle Furnaces with MoSi2 Heating Elements
  • Low thermal mass insulation for low external housing temperature
  • Built with low density ceramic bricks and ceramic fiber for faster heating rate
  • Double insulation includes air chamber lets work at high temperature continuously with air circulation
  • Programmable controller with multiple set points for ramp and dwell options
Vertical lift door keeps hot surface away when opening. Design is compact and lightweight having maximum operating temperature up to 1900 ºC with rapid heating through Kanthal® Super molybdenum disilicide (MoSi2) heating elements. Modern design metal case with chrome-phosphatizing base protection and external finish with heat-resistant metal paint. Heat resistance in refractory insulation of very low thermal conductivity coefficient. Door system adjusted on the furnace frame by pressure, allowing for complete sealing. Electrically and thermally insulated door handle. Rear ventilation via ceramic chimney.
These furnaces have lower front control panel, solid state relay, general safety switch and general safety contactor. Each model features PAD Digital control with microprocessor-based temperature controls, PID parameters, non-volatile memory and alarm. Different control options with multiple programming and has data logger and programmer communication by Ethernet/RS-232. These furnaces feature safety shut-off for thermocouple failure and door opening. Different range of accessories available as per the application need.
2,450,000.00 - ₹7,998,000.00INR / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Muffle Furnace, 3 L, 1650 C, MoSi2 Heating element; 1/Ea
₹2,450,000.00INR / Each
COMPARE
Muffle Furnace, 4 L, 1650 C, MoSi2 Heating element; 1/Ea
₹2,691,000.00INR / Each
COMPARE
Muffle Furnace, 6 L, 1650 C, MoSi2 Heating element; 1/Ea
₹2,968,000.00INR / Each
COMPARE
Muffle Furnace, 8 L, 1650 C, MoSi2 Heating element; 1/Ea
₹3,436,000.00INR / Each
COMPARE
Muffle Furnace, 12 L, 1650 C, MoSi2 Heating element; 1/Ea
₹4,007,000.00INR / Each
COMPARE
Muffle Furnace, 15 L, 1650 C, MoSi2 Heating element; 1/Ea
₹3,747,000.00INR / Each
COMPARE
Muffle Furnace, 3 L, 1800 C, MoSi2 Heating element; 1/Ea
₹2,963,000.00INR / Each
COMPARE
Muffle Furnace, 4 L, 1800 C, MoSi2 Heating element; 1/Ea
₹3,513,000.00INR / Each
COMPARE
Muffle Furnace, 6 L, 1800 C, MoSi2 Heating element; 1/Ea
₹3,546,000.00INR / Each
COMPARE
Muffle Furnace, 8 L, 1800 C, MoSi2 Heating element; 1/Ea
₹4,142,000.00INR / Each
COMPARE
Muffle Furnace, 12 L, 1800 C, MoSi2 Heating element; 1/Ea
₹4,452,000.00INR / Each
COMPARE
Muffle Furnace, 15 L, 1800 C, MoSi2 Heating element; 1/Ea
₹4,701,000.00INR / Each
COMPARE
Muffle Furnace, 3.4 L, 1900 C, MoSi2 Heating element; 1/Ea
₹3,999,000.00INR / Each
COMPARE
Muffle Furnace, 4.5 L, 1900 C, MoSi2 Heating element; 1/Ea
₹5,015,000.00INR / Each
COMPARE
Muffle Furnace, 6 L, 1900 C, MoSi2 Heating element; 1/Ea
₹5,408,000.00INR / Each
COMPARE
Muffle Furnace, 8 L, 1900 C, MoSi2 Heating element; 1/Ea
₹5,642,000.00INR / Each
COMPARE
Muffle Furnace, 12 L, 1900 C, MoSi2 Heating element; 1/Ea
₹6,640,000.00INR / Each
COMPARE
Muffle Furnace, 15 L, 1900 C, MoSi2 Heating element; 1/Ea
₹7,340,000.00INR / Each
COMPARE
Muffle Furnace, 22 L, 1900 C, MoSi2 Heating element; 1/Ea
₹7,998,000.00INR / Each
Showing 1- 19 of 19