Cole-Parmer® Muffle Furnaces with drop down & side swing door and ceramic fiber insulation
Designed for occasional intermittent work
  • Drop down door can be used as a work platform and side swing door is for user safety
  • Double Low thermal mass insulation for low external housing temperature
  • Built with insulating ceramic fiber for faster heating rate
Design is compact and lightweight, and consists of 1 or 2 heating plates with Kanthal® AF. Metal case with chrome-phosphatizing base protection and external finish with heat-resistant metal paint. Easy loading drop down door and side swing door close hermetically. These furnaces are built with insulating ceramic fiber. Furnace has a ceramic chimney.
These furnaces have lower front or lateral control panel, solid state relay. Each model features PAD Digital control with microprocessor, PID parameters, non-volatile memory and alarm. Different control options with multiple programming and has data logger and programmer communication by Ethernet/RS-232. These furnaces feature safety shut-off for thermocouple failure. Different range of accessories available as per the application need.
327,000.00 - ₹439,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Muffle Furnace, 8 L, 1000 C, Drop down and side swing door; 1/Ea
₹439,000.00INR / Each
COMPARE
Muffle Furnace, 2 L, 1100 C, Drop down and side swing door; 1/Ea
₹327,000.00INR / Each
COMPARE
Muffle Furnace, 3.6 L, 1100 C, Drop down and side swing door; 1/Ea
₹357,000.00INR / Each
COMPARE
Muffle Furnace, 3.6 L, 1100 C, Drop down and side swing door with large height; 1/Ea
₹436,000.00INR / Each