Cole-Parmer® Mini-Coolers
Keeps temperature-sensitive samples cool during testing
  • Non-slip rubber feet
  • Gridded and keyed lids provide sample identification
These mini-coolers protect temperature-sensitive samples at the work bench or in the freezer. The polycarbonate constructed coolers are ideal for keeping PCR reagents, enzymes, and other biochemical reagents cool while working at the bench.
19,200.00 - ₹41,600.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (cm) Lowest Set Temperature (° C) Description
Availability Pricing
COMPARE
14.5 0 Mini-Cooler, 0°C, 0.5-2mL, 12-Place, Green
₹19,200.00INR / Each
COMPARE
25.5 0 Mini-Cooler, 0°C, 0.5-2mL, 32-Place, Green
₹35,600.00INR / Each
COMPARE
25.5 0 Mini-Cooler, 0°C, 0.5-2mL, Gel Lid, 32-Place, Green
₹41,100.00INR / Each
COMPARE
14.5 -20 Mini-Cooler, -20°C, 0.5-2mL, 12-Place, Blue
₹19,200.00INR / Each
COMPARE
25.5 -20 Mini-Cooler, -20°C, 0.5-2mL, 32-Place, Blue
₹35,600.00INR / Each
COMPARE
25.5 -20 Mini-Cooler, -20°C, 0.5-2mL, Gel Lid, 32-Place, Blue
₹41,600.00INR / Each