Cole-Parmer Mini-Cooler, -20°C, 0.5-2mL, 32-Place, Blue

Representative image only.

Cole-Parmer Mini-Cooler, -20°C, 0.5-2mL, 32-Place, Blue

Cole-Parmer | Item: 4400251

Keeps temperature-sensitive samples cool during testing

₹35,600.00INR / Each

₹35,600.00INR / Each

Buy It Together

Current Item

Cole-Parmer Mini-Cooler, -20°C, 0.5-2mL, 32-Place, Blue
Cole-Parmer Mini-Cooler, -20°C, 0.5-2mL, 32-Place, Blue
4400251

₹35,600.00INR / Each
+ =
Total Price 104100.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Includes

0.5 mL tube adapters and temperature-retention graph.

Key Features

  • Non-slip rubber feet
  • Gridded and keyed lids provide sample identification
  • Temperature retention -15 °C for 1 hours

More About this Item

These mini-coolers protect temperature-sensitive samples at the work bench or in the freezer. The polycarbonate constructed coolers are ideal for keeping PCR reagents, enzymes, and other biochemical reagents cool while working at the bench.

Specifications
  • Tubes Held32
  • Tube Size0.5 to 2.0 mL
  • Lowest Set Temperature (° C)-20
  • ColorBlue
  • Height (in)4 1/10
  • Length (in)9 19/32
  • Width (in)3 29/32
  • Width (cm)10
  • Height (cm)10.5
  • Length (cm)25.5
  • MaterialPolycarbonate(PC)
  • DescriptionMini-Cooler, -20°C, 0.5-2mL, 32-Place, Blue
Reviews
Q&A