Cole-Parmer® Microtube Labels
Use in temperatures ranging from –40 to 121°C (–40 to 249°F)
  • Adhere to most plastics, glass, and metals
  • Resistant to detergents, oils, solvents, and caustic agents
These labels have adhesive that is designed to withstand extreme cold temperatures from –40 to 249°F (–40 to 121°C). Labels adhere to most plastics, glass, and metals. Resistant to detergents, oils, solvents, and caustic agents without peeling or falling off. Available in an array of colors and sizes the labels can be printed on and will not bleed. The laser printer labels come in stay-flat sheets that will not jam in laser printers.
4,000.00 - ₹23,800.00INR / Roll of 1
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Length (in) Width (in)
Availability Pricing
COMPARE
White 0.8125 0.2812
₹6,400.00INR / Roll of 1
COMPARE
White 1 0.5
₹4,200.00INR / Roll of 1
COMPARE
Red 1 0.5
₹4,200.00INR / Roll of 1
COMPARE
Yellow 1 0.5
₹7,100.00INR / Roll of 1
COMPARE
Green 1 0.5
₹4,200.00INR / Roll of 1
COMPARE
Blue 1 0.5
₹10,400.00INR / Roll of 1
COMPARE
White 1.25 0.5
₹17,000.00INR / Roll of 1
COMPARE
Green 1.25 0.5
₹6,000.00INR / Roll of 1
COMPARE
White 0.375 0.375
₹4,200.00INR / Roll of 1
COMPARE
Red 0.375 0.375
₹4,000.00INR / Roll of 1
COMPARE
Yellow 0.375 0.375
₹4,200.00INR / Roll of 1
COMPARE
Green 0.375 0.375
₹4,100.00INR / Roll of 1
COMPARE
Blue 0.375 0.375
₹4,200.00INR / Roll of 1
COMPARE
White 0.5 0.5
₹7,000.00INR / Roll of 1
COMPARE
Red 0.5 0.5
₹7,000.00INR / Roll of 1
COMPARE
Yellow 0.5 0.5
₹6,000.00INR / Roll of 1
COMPARE
Green 0.5 0.5
₹7,100.00INR / Roll of 1
COMPARE
Blue 0.5 0.5
₹6,000.00INR / Roll of 1
COMPARE
White
₹17,700.00INR / Pkg of 1000
COMPARE
White 1.25 0.5
₹17,000.00INR / Pkg of 1000
Showing 1 - 20 of 21