Cole-Parmer Microtube Circular Printer Labels, White, 3/8" Dia; 1000/Pk
Representative image only.
Cole-Parmer Microtube Circular Printer Labels, White, 3/8" Dia; 1000/Pk
Cole-Parmer | Item: 6220643
Use in temperatures ranging from –40 to 121°C (–40 to 249°F)
₹17,700.00INR / Pkg of 1000
₹17,700.00INR / Pkg of 1000
Product Description
Key Features
  • Adhere to most plastics, glass, and metals
  • Resistant to detergents, oils, solvents, and caustic agents
More About this Item
These labels have adhesive that is designed to withstand extreme cold temperatures from –40 to 249°F (–40 to 121°C). Labels adhere to most plastics, glass, and metals. Resistant to detergents, oils, solvents, and caustic agents without peeling or falling off. Available in an array of colors and sizes the labels can be printed on and will not bleed. The laser printer labels come in stay-flat sheets that will not jam in laser printers.
Specifications
  • ColorWhite
  • Diameter (in)0.375
  • DescriptionMicrotube Circular Printer Labels, White, 3/8" Dia; 1000/Pk