Cole-Parmer® Microscope Slide Cover Glasses, Soda Lime Glass
Specially treated to avoid sticking and breakage
  • No. 1 thickness (0.13 to 0.17 mm)
  • Comes in a hinged box
Slide coverglasses are ideal for manual use and automatic coverslippers.
220.00 - ₹270.00INR / Pkg of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Size Description
Availability Pricing
COMPARE
Square (18 x 18 mm) Cover Glasses, 18 x 18 mm; 100/PK
₹220.00INR / Pkg of 100
COMPARE
Square (20 x 20 mm) Cover Glasses, 20 x 20 mm; 100/PK
₹240.00INR / Pkg of 100
COMPARE
Square (22 x 22 mm) Cover Glasses, 22 x 22 mm; 100/PK
₹270.00INR / Pkg of 100