Cole-Parmer MicroPette Single-Channel Fixed Volume Pipettors
High quality pipettors at a value price!
  • Lightweight, ergonomic design
  • Easy to calibrate and maintain
These pipettors are affordable yet are manufactured from the latest materials to ensure a lightweight and durable laboratory tool that will provide years of reliable service. Each pipettor is factory-tested according to ISO 8655/DIN 12650 gravimetric standards with distilled water at 22° C.
A thumb controlled tip ejector allows convenient one-handed operation. The finger support keeps the instrument in place with minimum user effort. The ejector collar and tip cone can be removed for easy cleaning and maintenance. Tip cone is manufactured of chemical resistant materials and has built-in filter to protect the internal mechanism from contamination.
Call For Price
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy (%) Max Volume (mL) Max Volume (µL)
Availability Pricing
COMPARE
0.005 5
Call For Price
COMPARE
0.01 10
Call For Price
COMPARE
0.02 20
Call For Price
COMPARE
0.025 25
Call For Price
COMPARE
0.05 50
Call For Price
COMPARE
0.1 100
Call For Price
COMPARE
0.2 200
Call For Price
COMPARE
0.25 250
Call For Price
COMPARE
0.5 500
Call For Price
COMPARE
1 1000
Call For Price
COMPARE
2 2000
Call For Price
COMPARE
5 5000
Call For Price
Showing 1- 12 of 12