Cole-Parmer Pipettor Fixed Volume, 100uL
Representative image only.
Cole-Parmer Pipettor Fixed Volume, 100uL
Cole-Parmer | Item: 9470391
High quality pipettors at a value price!
Call For Price
for an additional
₹14,500.00INR / Each
for an additional
₹25,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Cole-Parmer Pipettor Fixed Volume, 100uL
Cole-Parmer Pipettor Fixed Volume, 100uL
9470391
Call For Price
+ =
Total Price 14500.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Lightweight, ergonomic design
  • Easy to calibrate and maintain
  • 3 year warranty
More About this Item
These pipettors are affordable yet are manufactured from the latest materials to ensure a lightweight and durable laboratory tool that will provide years of reliable service. Each pipettor is factory-tested according to ISO 8655/DIN 12650 gravimetric standards with distilled water at 22° C.
A thumb controlled tip ejector allows convenient one-handed operation. The finger support keeps the instrument in place with minimum user effort. The ejector collar and tip cone can be removed for easy cleaning and maintenance. Tip cone is manufactured of chemical resistant materials and has built-in filter to protect the internal mechanism from contamination.
3 Year Warranty
Specifications
  • SterileNo
  • Max Volume (µL)100
  • Max Volume (mL)0.1
  • Max Dispensing (mL)0.1
  • Tip EjectorYes
  • DescriptionPipettor Fixed Volume, 100uL