Cole-Parmer MicroPette Single-Channel Adjustable Volume Pipettors
High quality pipettors at a value price!
  • Lightweight, ergonomic design
  • Easy to calibrate and maintain
These pipettors are affordable yet are manufactured from the latest materials to ensure a lightweight and durable laboratory tool that will provide years of reliable service. Each pipettor is factory-tested according to ISO 8655/DIN 12650 gravimetric standards with distilled water at 22° C.
A thumb controlled tip ejector allows convenient one-handed operation. The finger support keeps the instrument in place with minimum user effort. The ejector collar and tip cone can be removed for easy cleaning and maintenance. Tip cone is manufactured of chemical resistant materials and has built-in filter to protect the internal mechanism from contamination.
The adjustable volume models have a digital display for clear volume setting.
Call For Price
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Autoclavable Max Volume (mL) Min Volume (mL)
Availability Pricing
COMPARE
0.0025 0.0001
Call For Price
COMPARE
0.01 0.0005
Call For Price
COMPARE
0.02 0.002
Call For Price
COMPARE
0.05 0.005
Call For Price
COMPARE
0.1 0.01
Call For Price
COMPARE
0.2 0.02
Call For Price
COMPARE
0.2 0.05
Call For Price
COMPARE
1 0.1
Call For Price
COMPARE
1 0.2
Call For Price
COMPARE
5 1
Call For Price
Showing 1- 10 of 10