Cole-Parmer® Kynar PVDF Gas Sampling Bags
Corrosive-resistant Kynar for all your application needs
  • Strong seams for added durability
All standard models have either an on/off valve (3⁄16'' nickel-plated brass hose barb) or a silicone septum in polypropylene housing; dual-valve models have both. Max temp: 160°F (71°C).
Standard Kynar Bags
Single-wide seam; available in 2-mil and 4-mil thick styles.
Dual-Valve Kynar Bags
Double-wide seamed bags have both on/off and septum valves for convenient mixing.
7,500.00 - ₹21,900.00INR / Each
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Width (in) Valve Volume (Liters)
Availability Pricing
COMPARE
6 On/off 0.3
₹7,500.00INR / Each
COMPARE
9 On/off 1.6
₹10,100.00INR / Each
COMPARE
12 On/off 3.8
₹11,100.00INR / Each
COMPARE
15 On/off 8.1
₹13,000.00INR / Each
COMPARE
18 On/off 20.3
₹13,000.00INR / Each
COMPARE
24 On/off 37.7
₹15,400.00INR / Each
COMPARE
30 On/off 85.7
₹19,700.00INR / Each
COMPARE
6 Septum 0.3
₹7,700.00INR / Each
COMPARE
9 Septum 1.6
₹7,700.00INR / Each
COMPARE
12 Septum 3.8
₹9,000.00INR / Each
COMPARE
18 Septum 20.3
₹11,700.00INR / Each
COMPARE
24 Septum 37.7
₹14,200.00INR / Each
COMPARE
30 Septum 85.7
₹18,500.00INR / Each
COMPARE
6 Dual valve, on/off and septum 0.3
₹9,300.00INR / Each
COMPARE
9 Dual valve, on/off and septum 1.6
₹10,900.00INR / Each
COMPARE
12 Dual valve, on/off and septum 3.8
₹12,100.00INR / Each
COMPARE
15 Dual valve, on/off and septum 8.1
₹15,300.00INR / Each
COMPARE
18 Dual valve, on/off and septum 20.3
₹15,100.00INR / Each
COMPARE
24 Dual valve, on/off and septum 37.7
₹17,400.00INR / Each
COMPARE
30 Dual valve, on/off and septum 85.7
₹21,900.00INR / Each
Showing 1- 20 of 20