Cole-Parmer® INC-400 Series Gravity Convection Incubators
Gravity convection provides a lower chance of sample cross-contamination
  • PID technology for high precision control and small temperature fluctuation
  • Extended-life silicone rubber gasket provides a long service life
  • Stainless steel inner chambers resists corrosion and is easy to clean
  • Antiskid shelf design is easy to operate with a single hand
  • Glass doors ensure perfect observation of the samples
Gravity convection allows heat to fill the chamber naturally, providing a lower chance of cross contamination between samples. These gravity convection incubators have a temperature range from ambient +5℃ to 65℃. The inner chamber is constructed of stainless steel with a round, curved inner angle for quick cleaning. The antiskid shelf design is easy to use with a single hand. Smaller models have a large outer glass door while larger models have an internal glass door. Glass doors avoid unnecessary heat dissipation and temperature changes when samples need to be viewed.
Easy-to-view LCD shows time, set temperature, actual temperature, running status, and fan status. Safety features include auto start after power loss/return, temperature deviation alarm, overtemperature set point alarm, and overcurrent protection.
113,000.00 - ₹454,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Liters) Max Temperature (° C) Power (VAC)
Availability Pricing
COMPARE
16 65 120
₹113,000.00INR / Each
COMPARE
16 65 220
₹222,000.00INR / Each
COMPARE
35 65 120
₹120,000.00INR / Each
COMPARE
35 65 220
₹235,000.00INR / Each
COMPARE
55 65 120
₹154,000.00INR / Each
COMPARE
55 65 220
₹302,000.00INR / Each
COMPARE
115 65 120
₹205,000.00INR / Each
COMPARE
115 65 220
₹401,000.00INR / Each
COMPARE
210 65 120
₹247,000.00INR / Each
COMPARE
210 65 220
₹454,000.00INR / Each
Showing 1- 10 of 10