Cole-Parmer® HPLC Syringe Filters
High quality, economical price
  • Syringe filters come with plastic jar for convenient storage
These nonsterile syringe filters are tested for physical properties and UV detectable extractables. Each unit features secure, enhanced female Luer-Lok™ inlet and male luer slip outlet, and solvent-resistant low extractable polypropylene housing. Select membranes available with 1.0 µm binder-free glass prefilter for high-solids sample. All syringe filters are autoclavable.
13,600.00 - ₹72,200.00INR / Pkg of 100
26 variations of this product are available.
See All Product Options Below
26 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Membrane Material Pore Size (µm)
Availability Pricing
COMPARE
4 Nylon 0.2
₹28,400.00INR / Pkg of 100
COMPARE
4 Nylon 0.45
₹33,000.00INR / Pkg of 100
COMPARE
17 Nylon 0.45
₹35,200.00INR / Pkg of 100
COMPARE
17 Nylon 0.2
₹33,400.00INR / Pkg of 100
COMPARE
30 Nylon 0.45
₹42,700.00INR / Pkg of 100
COMPARE
30 Nylon with Binder-free Glass Prefilter 0.45
₹49,400.00INR / Pkg of 100
COMPARE
30 Nylon 0.2
₹43,000.00INR / Pkg of 100
COMPARE
30 Nylon 1.5
₹44,600.00INR / Pkg of 100
COMPARE
30 Nylon 5
₹43,000.00INR / Pkg of 100
COMPARE
4 Polytetrafluoroethylene (PTFE) 0.45
₹47,700.00INR / Pkg of 100
COMPARE
4 Polytetrafluoroethylene (PTFE) 0.2
₹47,700.00INR / Pkg of 100
COMPARE
17 Polytetrafluoroethylene (PTFE) 0.45
₹43,000.00INR / Pkg of 100
COMPARE
17 Polytetrafluoroethylene (PTFE) 0.2
₹39,400.00INR / Pkg of 100
COMPARE
30 Polytetrafluoroethylene (PTFE) 0.45
₹44,500.00INR / Pkg of 100
COMPARE
30 Polytetrafluoroethylene (PTFE) 0.2
₹46,200.00INR / Pkg of 100
COMPARE
30 Polytetrafluoroethylene (PTFE) 1
₹45,500.00INR / Pkg of 100
COMPARE
17 Polyvinylidene fluoride (PVDF) 0.2
₹41,300.00INR / Pkg of 100
COMPARE
30 Polyvinylidene fluoride (PVDF) 0.2
₹52,900.00INR / Pkg of 100
COMPARE
17 Regenerated Cellulose (RC) 0.45
₹39,300.00INR / Pkg of 100
COMPARE
17 Polypropylene (PP) 0.2
₹36,900.00INR / Pkg of 100
Showing 1 - 20 of 26