Cole-Parmer HPLC Syringe Filters, Non Sterile, RC, 4 mm, .2 µm, 100/pk
Representative image only.
Cole-Parmer HPLC Syringe Filters, Non Sterile, RC, 4 mm, .2 µm, 100/pk
Cole-Parmer | Item: 3281662
High quality, economical price
₹87,700.00INR / Pkg
₹87,700.00INR / Pkg
Product Description
Key Features
  • Syringe filters come with plastic jar for convenient storage
More About this Item
These nonsterile syringe filters are tested for physical properties and UV detectable extractables. Each unit features secure, enhanced female Luer-Lok™ inlet and male luer slip outlet, and solvent-resistant low extractable polypropylene housing. Select membranes available with 1.0 µm binder-free glass prefilter for high-solids sample. All syringe filters are autoclavable.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
  • Membrane MaterialRegenerated Cellulose (RC)
  • Housing MaterialPolypropylene (PP)
  • DescriptionHPLC Syringe Filters, Non Sterile, RC, 4 mm, .2 µm, 100/pk