Cole-Parmer® Hose Barbs Fittings, Tee Reducer, Natural Polycarbonate
Two large barbs and one small barb to easily combine or split fluid flow
  • Sharp back-barb securely holds tubing in place
  • Transparent fittiings—easily view fluid flow
  • Meets FDA 21 CFR 177.1580 and NSF 51 requirements, and RoHS compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
Easily combine or split fluid flow using tee reducers with two large hose barbs and one small hose barb. A specially designed sharp back-barb expands tubing 40 to 50% above its normal ID for a leak-free, clamp-free seal up to 125 psi. Transparent fittings make it easy to verify fluid flow.
1,400.00 - ₹1,500.00INR / Pkg of 10
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Barb Size / Tubing Id (Port 3) (in)
Availability Pricing
COMPARE
1/8 1/8 1/16
₹1,400.00INR / Pkg of 10
COMPARE
1/8 1/8 3/32
₹1,400.00INR / Pkg of 10
COMPARE
5/32 5/32 1/16
₹1,400.00INR / Pkg of 10
COMPARE
5/32 5/32 1/8
₹1,400.00INR / Pkg of 10
COMPARE
5/32 5/32 3/32
₹1,400.00INR / Pkg of 10
COMPARE
3/16 3/16 1/8
₹1,400.00INR / Pkg of 10
COMPARE
1/4 1/4 1/8
₹1,500.00INR / Pkg of 10
COMPARE
1/4 1/4 3/32
₹1,500.00INR / Pkg of 10
Showing 1- 8 of 8