Cole-Parmer Hose Barb Fitting, Tee Reducer, Natural Polycarbonate, 5/32" ID x 5/32" ID x 3/32" ID; 10/Pk
Representative image only.
Cole-Parmer Hose Barb Fitting, Tee Reducer, Natural Polycarbonate, 5/32" ID x 5/32" ID x 3/32" ID; 10/Pk
Cole-Parmer | Item: 5014030
Two large barbs and one small barb to easily combine or split fluid flow
₹1,400.00INR / Pkg of 10
₹1,400.00INR / Pkg of 10
Product Description
Key Features
  • Sharp back-barb securely holds tubing in place
  • Transparent fittiings—easily view fluid flow
  • Meets FDA 21 CFR 177.1580 and NSF 51 requirements, and RoHS compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • 5/32" x 5/32" x 3/32" hose barbs
  • Manufacturer defect warranty
More About this Item
Easily combine or split fluid flow using tee reducers with two large hose barbs and one small hose barb. A specially designed sharp back-barb expands tubing 40 to 50% above its normal ID for a leak-free, clamp-free seal up to 125 psi. Transparent fittings make it easy to verify fluid flow.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
ISO 9001:2008 Certified Supplier
Specifications
  • Fitting MaterialPolycarbonate
  • Sterilization MethodEtO or gamma irradiation
  • Connection TypeHose Barb Tee Reducer
  • Number Of Ports2
  • Barb Size / Tubing Id (Port 1) (in)5/32
  • Barb Size / Tubing Id (Port 2) (in)5/32
  • Barb Size / Tubing Id (Port 3) (in)3/32
  • Flow PatternTee
  • ColorNatural
  • Max Temperature (° F)797
  • Max Temperature (° C)425
  • Regulatory ApprovalsNSF-51;RoHS;FDA;Non-DEHP
  • DescriptionHose Barb Fitting, Tee Reducer, Natural Polycarbonate, 5/32" x 5/32" x 3/32" Hose Barbs; 10/Pk
  • WarrantyManufacturer defect