Cole-Parmer® High-Purity Gas Regulators for Research
Designed for laboratory research using mildly corrosive gases
  • Diaphragm packless valve promotes system purity
  • Threadless seat design provides longer life
These stainless steel one- and two-stage high-purity regulators are designed for non-corrosive analytical, mildly corrossive and process applications requiring precise, stable delivery pressure control. A diaphragm packless outlet valve with a 1/4" compression tube fitting allows flow control and maintains system purity.
Single-stage regulators feature a convoluted stainless steel diaphragm seal that provides excellent regulating characteristics and allows for internal vacuum purging
Dual-stage regulators feature a a convoluted Hastelloy® C-22 diaphragm seal that prevents contamination by eliminating the need for a soft seal and provides a leak-rate design of less than 2 x 10–8 ccs helium. The two-stage models yield a delivery pressure change of less than 0.04/100 psi inlet change.
297,000.00 - ₹524,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Max Exhaust Flow Rate (SCFM) Max Supply Pressure (PSI)
Availability Pricing
COMPARE
₹297,000.00INR / Each
COMPARE
₹297,000.00INR / Each
COMPARE
₹524,000.00INR / Each
COMPARE
₹338,000.00INR / Each
COMPARE
₹338,000.00INR / Each
COMPARE
₹338,000.00INR / Each
COMPARE
₹338,000.00INR / Each
Showing 1- 7 of 7