Cole-Parmer Laboratory Gas Regulator, Dual Stage 590 CGA SKU 9820286
Representative image only.
Cole-Parmer Laboratory Gas Regulator, Dual Stage 590 CGA SKU 9820286
Cole-Parmer | Item: 9820286
Designed for laboratory research using mildly corrosive gases
₹338,000.00INR / Each
₹338,000.00INR / Each
Product Description
Key Features
  • Diaphragm packless valve promotes system purity
  • Threadless seat design provides longer life
  • 1 year warranty
More About this Item
These stainless steel one- and two-stage high-purity regulators are designed for non-corrosive analytical, mildly corrossive and process applications requiring precise, stable delivery pressure control. A diaphragm packless outlet valve with a 1/4" compression tube fitting allows flow control and maintains system purity.
Single-stage regulators feature a convoluted stainless steel diaphragm seal that provides excellent regulating characteristics and allows for internal vacuum purging
Dual-stage regulators feature a a convoluted Hastelloy® C-22 diaphragm seal that prevents contamination by eliminating the need for a soft seal and provides a leak-rate design of less than 2 x 10–8 ccs helium. The two-stage models yield a delivery pressure change of less than 0.04/100 psi inlet change.
1 Year Warranty
Specifications
  • Max Delivery Pressure (psig)60
  • Max Exhaust Flow Rate (SCFM)4.5
  • Max Inlet Pressure (PSI)4000
  • Outlet Connections¼" OD compression
  • CGA Fitting590
  • Min Operating Temperature (° F)-40
  • Max Operating Temperature (° F)165
  • Min Operating Temperature (° C)-40
  • Max Operating Temperature (° C)74
  • Max Supply Pressure (PSI)4000
  • Capacity (SCFH)270
  • Materials of Construction316 SS body & filter; SS gauges; Ni-plated brass bonnet; PCTFE® seat; Elgiloy diaphragm; 316 SS outlet valve
  • DescriptionLaboratory Gas Regulator, Dual Stage 590 CGA