Cole-Parmer® High-Form Crucibles, Nickel
Constructed from one piece of 24 gauge (0.025'') nickel, these crucibles are polished inside and out. Withstand temperatures to 1472°F (800°C).
7,000.00 - ₹16,100.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Overall Height (mm) Capacity (mL) Top Outside Diameter (mm)
Availability Pricing
COMPARE
31 15 31
₹7,000.00INR / Each
COMPARE
35 25 35
₹7,000.00INR / Each
COMPARE
45 50 45
₹9,700.00INR / Each
COMPARE
56 100 56
₹16,100.00INR / Each