Cole-Parmer® High-Capacity Vortex Mixer
A true vortex in every vessel
  • Mix more samples at a time—large, heavy-duty platform
  • Choose from constant or pulse mixing settings—run up to 100 hours
  • Accepts almost any tube rack—adjustable
These versatile vortex mixer/shakers are ideal for medium to high throughput laboratories. The large 7" x 12" (17.8 x 30.5 cm) platform accepts standard racks for tubes, vials, flasks, cylinders, and microwell plates, or select from our pre-drilled foam rack options.
Mixers feature a balanced drive and oversized motor to deliver accurate and reproducible speeds, up to 2500 rpm, with minimal vibration. Microprocessor control provides a programmable pulsing mode for time specific interval mixing and optimal sample agitation. Set the run time and operate unattended for up to 100 hours.
Available in standard or QuEChERS versions—QuEChERS model (04724-01) includes a rack starter kit.
666,000.00 - ₹679,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Power (VAC) Description
Availability Pricing
COMPARE
2500 115 / 230 High-Capacity Vortex Mixer, QuEChERS; 115/230 VAC
₹666,000.00INR / Each
COMPARE
2500 115 / 230 High-Capacity Vortex Mixer, Standard; 115/230 VAC
₹679,000.00INR / Each