Cole-Parmer® High Accuracy Miniature Digital Gauge
Two styles of gauges feature 1% full-scale accuracy
  • Brass center back-mount process connections
  • Single-button function for on/units/off
  • Five-minute auto power-off preserves battery life
Designed for use in numerous applications and industries, these digital gauges can be used for air pressure measurement in marine and diving, filtration, compressor and pneumatic regulators measurement, laboratory vacuum and pressure measurement, as well as aviation and defense, and general OEM use where air or compatible gases are used and brass can be used.
These high-accuracy, low-cost digital gauges have a four-digit, 1/2"H LCD and read in psi, bar, mPa, and kg/cm2. Gauges have a 1 3/4" (44.5 mm) dial and come in three styles — 0 to 145 psi in 1/8" or 1/4" NPT(M) connection, and vacuum to 100 psi in 1/8" NPT(M) connection.
10,700.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Process Connection
Availability Pricing
COMPARE
±1% full-scale 100 1/8" NPT(M)
Call For Price
COMPARE
±1% full-scale 145 1/8" NPT(M)
₹10,700.00INR / Each
COMPARE
±1% full-scale 145 1/4" NPT(M)
₹10,700.00INR / Each