Cole-Parmer® Heavy-Walled Wide Mouth Carboys, HDPE
Heavy wall for added durability—ship without worry!
  • Chemical resistant
  • Handle allows for easy carrying of solutions
Economical, durable carboys are resistant to most chemicals. Heavy-duty swing handle allows easy carrying for your liquids and powders. Features LDPE neck insert plug for maximum security.
7,900.00 - ₹11,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
COMPARE
5 90 mm High Density Polyethyene (HDPE)
₹7,900.00INR / Each
COMPARE
10 120 mm High Density Polyethyene (HDPE)
₹11,700.00INR / Each