Cole-Parmer® Heavy-Duty Three-Prong Utility Clamps
Securely hold your flasks, beakers and burettes
  • An additional holder must be purchased in order to attach clamps to lab-frames, support stands or other apparatus
Featuring an innovative closed yoke design that minimizes contamination and corrosion of internal components and is easier to clean and decontaminate. This design allows use of the clamp’s full depth – no springs or internal workings to get in the way. Choose single adjustment style for standard use where one prong or “finger” is adjusted using a wing nut, while the other prong remains stationary or for a more versatile clamp choose the dual adjustment style. Utilizing thumbscrews, both prongs are easily adjusted over a wider range. Available in 3 sizes, small, medium, and large. Constructed of nickel plated zinc. Supplied with non-slip vinyl sleeves or for temperatures above 212°F (100°C), fiberglass covers. The prong height for these clamps make them ideal for use in holding and securing glassware with precision ground glass joints. The chart details which Heavy-Duty clamps are recommended:
Clamp Joint Size (mm)
Small Dual Adjust 10/30, 12/30, 14/20, 19/22, 24/40, 29/42, 34/45
Medium Dual Adjust 14/40, 19/38
Large Dual Adjust 45/50
8,400.00 - ₹11,600.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Clamp Material Clamp Type Length (in)
Availability Pricing
COMPARE
Nickel-plated Zinc with red vinyl and black fiberglass sleeves Equipment, Two- or Three-Prong 10.25
₹10,100.00INR / Each
COMPARE
Nickel-plated Zinc with red vinyl and black fiberglass sleeves Equipment, Two- or Three-Prong 9.02
₹9,100.00INR / Each
COMPARE
Nickel-plated Zinc with red vinyl and black fiberglass sleeves Equipment, Two- or Three-Prong 6.39
₹8,900.00INR / Each
COMPARE
Nickel-plated Zinc with red vinyl and black fiberglass sleeves Equipment, Two- or Three-Prong 5.98
₹8,800.00INR / Each
COMPARE
Nickel-plated Zinc with red vinyl and black fiberglass sleeves Equipment, Two- or Three-Prong 8.75
₹8,400.00INR / Each
COMPARE
Nickel-plated Zinc with red vinyl and black fiberglass sleeves Equipment, Two- or Three-Prong 10.25
₹11,600.00INR / Each