Cole-Parmer® Glass Vessels for PTFE Tissue Grinders
Glass vessels for use with Cole-Parmer plain or serrated PTFE head tissue grinders
  • Borosilicate glass
  • Use with matching tissue grinder
Borosilicate glass vessels are intended for use with Cole-Parmer plain or serrated PTFE head tissue grinders, sold separately. The pair effectively homogenizes a variety of tissue samples. When properly matched, the clearance between plunger and glass is 0.15 to 0.25 mm. Volumes stated are working volumes with the plunger in place.
14,200.00 - ₹28,600.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Liters) Capacity (mL) Material
Availability Pricing
COMPARE
0.002 2 Borosilicate glass
₹14,200.00INR / Each
COMPARE
0.005 5 Borosilicate glass
₹16,500.00INR / Each
COMPARE
0.01 10 Borosilicate glass
₹16,900.00INR / Each
COMPARE
0.015 15 Borosilicate glass
₹19,600.00INR / Each
COMPARE
0.03 30 Borosilicate glass
₹23,100.00INR / Each
COMPARE
0.05 50 Borosilicate glass
₹28,600.00INR / Each