Cole-Parmer® Glass Dome Desiccators
Clear glass provides excellent visibility
  • Remove cover easily with built-in knob
  • Autoclavable
Manufactured from durable borosilicate glass, these clear glass dome desiccators are designed for analytical work. The borosilicate glass lid features a molded-in knob for easy handling of the cover. To create an airtight seal, it is recommended you grease the rim and lid of the desiccator.
The included perforated porcelain desiccator plate fits onto a ridge above the lower chamber. A standard drying agent such as silica or anhydrous calcium chloride can be placed in the lower chamber.
30,800.00 - ₹92,100.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Clearance Above Plate (mm) Exterior Height (InchShort) Flange Outside Diameter (OD) (cm)
Availability Pricing
COMPARE
130 9 15
₹30,800.00INR / Each
COMPARE
180 11 20
₹50,700.00INR / Each
COMPARE
215 5 30
₹92,100.00INR / Each