Cole-Parmer® Gel-Filled, Side-Mount pH Electrodes
Mount these versatile electrodes in any position
  • Durable—ideal for your demanding applications
Sealed AgCl reference cell requires no refilling during use. Rugged PVDF housing with 1/2'' NPT(M) thread withstands pressures up 100 psi, good for measurements up to 14 pH units. Includes a 10-ft cable with BNC connector.
27,100.00 - ₹42,100.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
gel-filled side-mount electrode, single-junction, 10-ft cable, BNC
₹27,100.00INR / Each
COMPARE
Gel-Filled Side-Mount pH Probe, SJ/PVDF/100Ohm RTD; Plug/BNC
₹35,600.00INR / Each
COMPARE
Gel-Filled Side-Mount pH Probe, DJ/PVDF/100hm RTD; Tinned/BNC
₹42,100.00INR / Each