Cole-Parmer Gel-Filled Side-Mount pH Probe, DJ/PVDF/100hm RTD; Tinned/BNC
Representative image only.
Cole-Parmer Gel-Filled Side-Mount pH Probe, DJ/PVDF/100hm RTD; Tinned/BNC
Cole-Parmer | Item: 2701130
Mount these versatile electrodes in any position
₹42,100.00INR / Each
₹42,100.00INR / Each
Product Description
Key Features
  • Durable—ideal for your demanding applications
  • 1 year warranty
More About this Item
Sealed AgCl reference cell requires no refilling during use. Rugged PVDF housing with 1/2'' NPT(M) thread withstands pressures up 100 psi, good for measurements up to 14 pH units. Includes a 10-ft cable with BNC connector.
1 Year Warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialPVDF
  • Min pH (pH)1
  • Max pH (pH)14
  • Max Temperature (° C)100
  • Temperature CompensationYes
  • Thread Size NPT(M) (in)1/2
  • ATC ConnectorTinned ends
  • ATC Element100 Ω Pt RTD
  • ConnectorBNC
  • Cable Length (m)3
  • DescriptionGel-Filled Side-Mount pH Probe, DJ/PVDF/100hm RTD; Tinned/BNC