Cole-Parmer® Fume Hood Cabinets
Provides additional storage and safety supports fume hoods
  • Chemical resistant finish
  • Ideal for labs with limited space
These two door fume hood cabinets feature all welded steel construction with white chemical resistant finish. Dimensions are 35” high and 22” deep. Available in standard base and acid storage, the cabinets come in 30”, 36”, and 40” sizes.
The interior of the acid cabinets is lined with chemical resistant panels and sealed, ideal for acid and corrosive chemicals. The molded one-piece fiberglass liner is easy to clean.
560,000.00 - ₹1,397,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (in) Hood Width (in) Description
Availability Pricing
COMPARE
35 30 Acid-storage base cabinet, 30" W
₹680,000.00INR / Each
COMPARE
35 30 Base Cabinet for Fume Hoods, 30" W
₹560,000.00INR / Each
COMPARE
35 36 Acid-storage base cabinet, 36" W
₹1,397,000.00INR / Each
COMPARE
35 36 Base Cabinet for Fume Hoods, 36" W
₹674,000.00INR / Each
COMPARE
35 48 Acid-storage base cabinet, 48" W
₹727,000.00INR / Each
COMPARE
35 48 Base Cabinet for Fume Hoods, 48" W
₹803,000.00INR / Each