Cole-Parmer® Fluorescent Red Dye Tracers
Determine water flow in closed circuits or open channels
  • Biodegradable and nontoxic
Dyes diffuse rapidly in water, producing a vivid, easy-to-see color. Excellent for identifying diffusion and dispersion patterns, rates of flow, discharge locations, seepage areas, sea marking, and more.
Fluorescent red dye (rhodamine WT) is noncarcinogenic. Ideal in yellowed water or water with heavy algae growth. Use an ultraviolet lamp (order separately) to trace fluorescent red dyes where normal visual detection is difficult. Minimum detectable concentration of all dyes in clear water is 1 ppm.
8,900.00 - ₹52,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Fluorescent Fwt Red Dye Tablets, 200 Tablets
₹13,300.00INR / Each
COMPARE
Fluorescent Fwt Red Dye Concentrate, Gallon
₹33,400.00INR / Gallon of 1
COMPARE
Fluorescent Fwt Red Dye Concentrate, Pint
₹8,900.00INR / Pint of 1
COMPARE
Fluorescent Fwt Red Tracer Dye Powder, 1 Lb
₹52,000.00INR / Each