Cole-Parmer Fluorescent Fwt Red Dye Concentrate, Pint
Representative image only.
Cole-Parmer Fluorescent Fwt Red Dye Concentrate, Pint
Cole-Parmer | Item: 0029816
Determine water flow in closed circuits or open channels
₹8,900.00INR / Pint of 1
₹8,900.00INR / Pint of 1
Product Description
Key Features
  • Biodegradable and nontoxic
More About this Item
Dyes diffuse rapidly in water, producing a vivid, easy-to-see color. Excellent for identifying diffusion and dispersion patterns, rates of flow, discharge locations, seepage areas, sea marking, and more.
Fluorescent red dye (rhodamine WT) is noncarcinogenic. Ideal in yellowed water or water with heavy algae growth. Use an ultraviolet lamp (order separately) to trace fluorescent red dyes where normal visual detection is difficult. Minimum detectable concentration of all dyes in clear water is 1 ppm.
NSF international certified
Specifications
  • Reagent PlatformLiquid concentrate
  • Gallons Of Water For 1 ppm (Gal)3125
  • DescriptionFluorescent Fwt Red Dye Concentrate, Pint