Cole-Parmer® Flock-Lined Nitrile Chemical-Resistant Gloves
High-performance nitrile formulation offers outstanding combination of strength and chemical resistance
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Outstanding protection from acids, caustics, solvent & chemicals Hand specific, ergonomic design with textured surface for excellent grip on wet & dry condition Flock-lined to prevent skin irritation Abrasion, cut and puncture resistant Available in 15 mil and 18 mil thickness Comply with US FDA CFR 21Part 177.2600 & EC regulation
The gloves offer very good abrasion and puncture resistance. The exclusive glove shape is optimized for the best possible fit on more than forty different dimensions. Choose the green nitrile for protection from fuels, grease, chemicals and solvents.
37,300.00 - ₹38,500.00INR / Case of 288
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Size Thickness (mil) Description
Availability Pricing
COMPARE
Small 18 Cole-Parmer CR GLOVE NITRILE 13" 18 mil Small, 144 pairs/Cs
₹38,500.00INR / Case of 288
COMPARE
Medium 18 Cole-Parmer CR GLOVE NITRILE 13" 18 mil Medium, 144 pairs/Cs
₹38,500.00INR / Case of 288
COMPARE
Large 18 Cole-Parmer CR GLOVE NITRILE 13" 18 mil Large, 144 pairs/Cs
₹38,500.00INR / Case of 288
COMPARE
X-Large 18 Cole-Parmer CR GLOVE NITRILE 13" 18 mil X-Large, 144 pairs/Cs
₹38,500.00INR / Case of 288
COMPARE
Small 15 Cole-Parmer CR GLOVE NITRILE 13" 15 mil Small, 144 pairs/Cs
₹37,300.00INR / Case of 288
COMPARE
Medium 15 Cole-Parmer CR GLOVE NITRILE 13" 15 mil Medium, 144 pairs/Cs
₹37,300.00INR / Case of 288
COMPARE
Large 15 Cole-Parmer CR GLOVE NITRILE 13" 15 mil Large, 144 pairs/Cs
₹37,300.00INR / Case of 288
COMPARE
X-Large 15 Cole-Parmer CR GLOVE NITRILE 13" 15 mil X-Large, 144 pairs/Cs
₹37,300.00INR / Case of 288
Showing 1- 8 of 8