Cole-Parmer® Extra-Thick PTFE Adhesive Tape
Extra-thick PTFE tape is available in unique thicknesses (20, 30, and 60 mil) with a high tack pressure-sensitive adhesive backing. PTFE offers a wide range of high performance characteristics—nonstick, low-friction surface; excellent chemical and temperature resistance; and excellent electrical insulation. PTFE tape has a peel-off liner and can be easily cut with scissors. Temperature range is –40 to 350°F ( 40 to 176°C).
15,900.00 - ₹101,000.00INR / Roll of 1
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Thickness (mil) Width (in)
Availability Pricing
COMPARE
15 20 1
₹15,900.00INR / Roll of 1
COMPARE
15 20 2
₹30,600.00INR / Roll of 1
COMPARE
15 30 1
₹22,100.00INR / Roll of 1
COMPARE
15 60 1
₹31,700.00INR / Roll of 1
COMPARE
15 60 2
₹101,000.00INR / Roll of 1