Cole-Parmer® Economical Square HDPE Carboys with Handles
Handle and pour contents easily
  • Durable, leakproof square carboys save space
  • Large carrying handles and an indented bottom make handling and pouring easy
  • Molded HDPE threaded closure included
  • Can be drilled to accept a 3/4'' faucet, sold separately
  • Temperature range: 68 to 230 °F (20 to 110 °C)
There is a molded-in handle and indented bottom for easy grabbing. No leaks with leakproof HDPE threaded cap. Dispensing can be made easier by drilling and attaching optional faucet. Withstand temperature from 68 to 248°F (20 to 120°C).
6,900.00 - ₹32,300.00INR / Case of 4
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
COMPARE
10 63 mm High Density Polyethyene (HDPE)
₹6,900.00INR / Each
COMPARE
20 70-400 thread 22mm High Density Polyethyene (HDPE)
₹7,400.00INR / Each
COMPARE
10 63 mm High Density Polyethyene (HDPE)
₹27,900.00INR / Case of 4
COMPARE
20 63 mm High Density Polyethyene (HDPE)
₹32,300.00INR / Case of 4