Cole-Parmer® Drying Rack and Accessories
SureGrip hook securely holds glassware in place to protect against breakage
  • Accommodates various glassware types
  • Angled pegs hold even large, heavy pieces securely in place
Use this versatile rack for drying various types of glassware. Built-in channels let you place pegs where you need to maximize the drying space. Mix and match small and large pegs to accommodate all glassware varieties. The rack is made of durable, easy-to-clean polystyrene and can handle up to 77 lbs of glassware. A unique drying basket features easily stores small items and channels to hold pipettes.
2,700.00 - ₹47,700.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Material Description
Availability Pricing
COMPARE
White Polystyrene Drying Rack Set, Wall/Bench Mount
₹47,700.00INR / Each
COMPARE
Clear ABS Drying Rack Replacement Basket, Clear; 2/PK
₹3,200.00INR / Pkg of 2
COMPARE
Green/Blue ABS Drying Rack Replacement Pegs; 10/PK
₹7,600.00INR / Pkg of 10
COMPARE
White Silicone Drying Rack Replacement Draining Set
₹2,700.00INR / Each