Cole-Parmer® Disposable 1-Way Stopcocks with Luer Fittings, Polycarbonate
Stopcocks provide accurate, leak-free flow control
  • Arrows indicate flow direction
  • Consistent inside diameter minimizes turbulence and bubble entrapment
  • Use with both liquids and gases
  • DEHP free
  • Polycarbonate (PC) housing and polyethylene rotor
Stopcocks provide an easy on/off control of your fluid. One-way stopcocks feature one female Luer fitting and a male Luer fitting with or without a lock. Luer fittings on stopcocks ensure a quick, leak-free connection. Handles indicate flow direction, enhancing operational efficiency and minimizing errors. Stopcocks are made of polyethylene rotor and transparent polycarbonate body, making it easy to see the flow.
2,700.00 - ₹22,200.00INR / Pkg of 10
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Connection Type Luer Connection Type (Port 2) Quantity
Availability Pricing
COMPARE
Female Luer x Male Locking Luer Male Locking Luer 10
₹3,000.00INR / Pkg of 10
COMPARE
Female Luer x Male Slip Luer Male Slip Luer 10
₹2,700.00INR / Pkg of 10
COMPARE
Female Luer x Male Locking Luer Male Locking Luer 100
₹21,100.00INR / Pkg of 100
COMPARE
Female Luer x Male Slip Luer Male Slip Luer 100
₹22,200.00INR / Pkg of 100