Cole-Parmer Disposable 1-Way Stopcock, PC, Female x Male Locking; 10/Pk
Representative image only.
Cole-Parmer Disposable 1-Way Stopcock, PC, Female x Male Locking; 10/Pk
Cole-Parmer | Item: 4852326
Stopcocks provide accurate, leak-free flow control
₹3,000.00INR / Pkg of 10
₹3,000.00INR / Pkg of 10
Product Description
Key Features
  • Arrows indicate flow direction
  • Consistent inside diameter minimizes turbulence and bubble entrapment
  • Use with both liquids and gases
  • DEHP free
  • Polycarbonate (PC) housing and polyethylene rotor
  • Female Luer x Male Locking Luer
  • Pack of 10
More About this Item
Stopcocks provide an easy on/off control of your fluid. One-way stopcocks feature one female Luer fitting and a male Luer fitting with or without a lock. Luer fittings on stopcocks ensure a quick, leak-free connection. Handles indicate flow direction, enhancing operational efficiency and minimizing errors. Stopcocks are made of polyethylene rotor and transparent polycarbonate body, making it easy to see the flow.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPolycarbonate
  • Sterilization MethodEtO
  • Connection TypeFemale Luer x Male Locking Luer
  • Number Of Ports2
  • Flow PatternStraight
  • ColorClear
  • Luer Connection Type (Port 1)Female Luer
  • Luer Connection Type (Port 2)Male Locking Luer
  • Quantity10
  • DescriptionDisposable 1-Way Stopcock, PC, Female x Male Locking; 10/Pk