Cole-Parmer® Deep-Well Microplates
Don't compromise your research
  • RNase, DNase and endotoxin free for higher purity and better results
  • Polypropylene plates are compatible with solvents and additives
  • Withstand temperatures from -80 to 120°C, ideal for both molecular and storage applications
These 96-well microplates are manufactured from 100% virgin polypropylene (PP), with a toughened option available for long-term storage. Select round-bottom plates for more efficient resuspension or V-bottom plates for precise sample recovery. Plates are RNase, DNase and endotoxin-free to ensure purity and improve results of PCR and other molecular techniques. Use with microplate sealing mats to reduce evaporation and contamination.
88,800.00INR / Pkg of 50
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Well Shape Well Volume (mL)
Availability Pricing
COMPARE
Round with round bottom 2
Call For Price
COMPARE
Round with round bottom 1
Call For Price
COMPARE
Square with V-shaped bottom 2
₹88,800.00INR / Pkg of 50
COMPARE
Square with V-shaped bottom 2