Cole-Parmer® Cryogenic Label for Laser Printers
Perfect for cryogenic storage — withstand temperatures from -321 to 302°F (-196 to 150°C).
  • Will not jam laser printers
  • Resistant to detergents, oils, solvents, and caustic agents without peeling or falling off
These labels, which will not jam laser printers will withstand conventional and cryogenic storage to -321°F (-196°C), boiling water baths to 212°F (100°C), and dry heat up to 302°F (150°C). Labels adhere to most plastics, glass, and metals.
10,500.00 - ₹13,100.00INR / Pkg of 1040
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Length (in) Width (in) Description
Availability Pricing
COMPARE
1 1/2 3/4 Cryogenic label for laser printers, 1 1/2"W x 3/4"H
₹12,900.00INR / Pkg of 12
COMPARE
1 11/16 3/4 Cryogenic label for laser printers, 1 11/16"W x 3/4"H
₹13,100.00INR / Pkg of 1040
COMPARE
1 9/32 1/2 Cryogenic Label for Laser Printers, 1 9/32" W x 1/2" H
₹10,500.00INR / Pkg of 1700