Cole-Parmer Cryogenic Label for Laser Printers, 1 9/32" W x 1/2" H
Representative image only.
Cole-Parmer Cryogenic Label for Laser Printers, 1 9/32" W x 1/2" H
Cole-Parmer | Item: 0653008
Perfect for cryogenic storage — withstand temperatures from -321 to 302°F (-196 to 150°C).
₹10,500.00INR / Pkg of 1700
₹10,500.00INR / Pkg of 1700
Product Description
Key Features
  • Will not jam laser printers
  • Resistant to detergents, oils, solvents, and caustic agents without peeling or falling off
More About this Item
These labels, which will not jam laser printers will withstand conventional and cryogenic storage to -321°F (-196°C), boiling water baths to 212°F (100°C), and dry heat up to 302°F (150°C). Labels adhere to most plastics, glass, and metals.
Specifications
  • Width (in)1/2
  • Length (in)1 9/32
  • DescriptionCryogenic Label for Laser Printers, 1 9/32" W x 1/2" H