Cole-Parmer® Essentials Crucible Lids, Nickel
Crucible lids are constructed from 24 gauge (0.025") nickel and withstand temperatures to 1472°F (800°C).
2,500.00 - ₹3,900.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL)
Availability Pricing
COMPARE
15
₹2,500.00INR / Each
COMPARE
25
₹3,100.00INR / Each
COMPARE
50
₹3,300.00INR / Each
COMPARE
100
₹3,900.00INR / Each