Cole-Parmer® Cooling Packs and Accessories
Transport your critical samples without worry
  • Maintain specific temperature ranges for extended periods of time for improved sample protection
  • Get the temperature protection you need with choice of durable transport bags or boxes
  • Modular, color-coded cooling packs allow flexible configurations for different-sized samples
  • Ensure sample integrity with optional security sealing ties
Cooling end-caps, cooling frames, and cooling blocks can be purchased individually or as part of a kit pairing nicely with selected bag or box to provide maximum protection. Use the cooling end-caps and cooling frames together to encapsulate the entire sample. This modular system can accommodate various sample sizes to provide a complete thermal barrier. Cooling packs come in standard 4 °C (color coded blue) and 22 °C (color coded green). Temperature ranges are 2 to 8 °C and 20 to 24 °C respectively; these ranges can be maintained up to 15 hours.
Cooling end-caps and cooling blocks include a temperature indicator label.
24,700.00 - ₹61,700.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Cooling End-caps, 22°C, Green, Pair of 2
₹61,700.00INR / Pair of 2
COMPARE
Cooling Frames, 22°C, Green Pair of 2
₹61,700.00INR / Pair of 2
COMPARE
Cooling End-caps, 4°C, Blue, Pair of 2
₹61,700.00INR / Pair of 2
COMPARE
Cooling Frames, 4°C, Blue, Pair of 2
₹61,700.00INR / Pair of 2
COMPARE
Cooling Block 22°C, Green, 1L
₹24,700.00INR / Each
COMPARE
Cooling Block Slim, 22°C, Green, 1L
₹24,700.00INR / Each
COMPARE
Cooling Block 4°C, Blue, 1L
₹26,400.00INR / Each
COMPARE
Cooling Block Slim, 4°C, Blue, 1L
₹26,400.00INR / Each
COMPARE
Cooling Block 4°C, Blue, 2L
₹57,000.00INR / Each
COMPARE
Security Sealing Ties, 100/cs
₹46,500.00INR / Case of 100
Showing 1- 10 of 10