Cole-Parmer Security Sealing Ties, 100/cs
Representative image only.
Cole-Parmer Security Sealing Ties, 100/cs
Cole-Parmer | Item: 0372619
Transport your critical samples without worry
₹46,500.00INR / Case of 100
₹46,500.00INR / Case of 100
Product Description
Key Features
  • Maintain specific temperature ranges for extended periods of time for improved sample protection
  • Get the temperature protection you need with choice of durable transport bags or boxes
  • Modular, color-coded cooling packs allow flexible configurations for different-sized samples
  • Ensure sample integrity with optional security sealing ties
  • Seals boxes to protect integrity
  • 1 year warranty
More About this Item
Cooling end-caps, cooling frames, and cooling blocks can be purchased individually or as part of a kit pairing nicely with selected bag or box to provide maximum protection. Use the cooling end-caps and cooling frames together to encapsulate the entire sample. This modular system can accommodate various sample sizes to provide a complete thermal barrier. Cooling packs come in standard 4 °C (color coded blue) and 22 °C (color coded green). Temperature ranges are 2 to 8 °C and 20 to 24 °C respectively; these ranges can be maintained up to 15 hours.
Cooling end-caps and cooling blocks include a temperature indicator label.
†Based on ambient temperature of 25°C. Estimates can vary depending on how samples are packed.
1 Year Warranty
Specifications
  • Description Security Sealing Ties, 100/cs