Cole-Parmer® Convertible Laser Tachometers
Rugged aluminum construction – ideal for industrial applications
  • Stores up to 13 readings
  • Accurately measure rpm from up to 165" (420 cm) away!
Select the LCD model for use in bright light or the LED model for easy reading in dim light. A laser spot makes these tachometers easy to aim. Use the memory button to recall last, maximum, minimum, and ten stored readings. Tachometers feature an overrange indicator and low battery indicator.
118,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Display Type Max Measuring Distance (in) Max RPM Range (rpm)
Availability Pricing
COMPARE
5-digit LCD, 0.5" H (1.3 cm) 165 99999
₹118,000.00INR / Each
COMPARE
5-digit LED, 0.5" H (1.3 cm) 165 99999
₹118,000.00INR / Each