Cole-Parmer® Clear Rain Gauges
No more guessing about rainfall
  • Easy to read rain level in English or Metric units
Measure accumulated precipitation with these clear buterate cylindrical rain gauges. Area ratio between knife-edge funnel and inner receiver is 10 to 1; inner receiver has molded-on graduations for easy reading. Gauges include buterate mounting bracket, screws, and a data recording sheet for logging rainfall data up to one year. Both models measure 14"L x 4.1" dia. Shpg wt 3 lb (1.4 kg).
11,100.00 - ₹11,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Rain Fall (in) Rain Fall Accuracy (in)
Availability Pricing
COMPARE
11 ±0.01"
₹11,200.00INR / Each
COMPARE
9.92 ±0.2 mm
₹11,100.00INR / Each