Cole-Parmer® Cleanroom Safety Shoes
Professional safety shoes with comfort, durability and protection
Features :
• Upper material: Buffalo embossed leather, thickness: 1.8 to 2.0 mm
• Steel toe: Conforms to ISO 20345:2011, Heel energy absorption: 3000 kgs or 20 joules
• Penetration resistance(steel midsole): Conforms to ISO 20345:2011 heavy work standards 23 kg/45 cm,1100 newtons
• Sole: Double density polyurethane outsole, with high resistance to slip (oil and slip resistant)
• Insole: Antistatic polyurethane insole
• Lining: made of Campela/ air mesh/ fur, with good absorption of sweat, can keep dry and comfortable
Conformity to Standards Conformity
• Feet protection Conforms to ISO 20345:2011. Under 23kg hammer toe drop height of 900 mm or 200J
• impact energy after impact, the toe internal clearance ≥ 15.0mm.
• Electrical isolation Conforms to ISO 20345:2011. With good isolation,
• Testing voltage: 6000V, Testing time: 1 min , Leakage current ≤1.8mA.
• Acid Resistance Conforms to GB12018-1989.
• Alkali Resistance Conforms to GB12018-1989.
• Oil resistance Conforms to GB21146-2007 FO.
• Antistatic property Conforms to ISO 20345:2011. Surface Resistance 1.3X106Ω
57,900.00 - ₹61,000.00INR / Case of 10
26 variations of this product are available.
See All Product Options Below
26 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Description
Availability Pricing
COMPARE
White Cleanroom Safety Shoe, White, Size -40; 10 Pairs/Cs
₹57,900.00INR / Case of 10
COMPARE
White Cleanroom Safety Shoe, White, Size -42; 10 Pairs/Cs
₹57,900.00INR / Case of 10
COMPARE
White Cleanroom Safety Shoe, White, Size -44; 10 Pairs/Cs
₹57,900.00INR / Case of 10
COMPARE
White Cleanroom Safety Shoe, White, Size -46; 10 Pairs/Cs
₹57,900.00INR / Case of 10
COMPARE
White Cleanroom Safety Shoe, White, Size 36; 10 Pairs/Cs
₹57,900.00INR / Case of 10
COMPARE
White Cleanroom Safety Shoe, White, Size 37; 10 Pairs/Cs
₹57,900.00INR / Case of 10
COMPARE
White Cleanroom Safety Shoe, White, Size 38; 10 Pairs/Cs
₹57,900.00INR / Case of 10
COMPARE
White Cleanroom Safety Shoe, White, Size 39; 10 Pairs/Cs
₹57,900.00INR / Case of 10
COMPARE
White Cleanroom Safety Shoe, White, Size 41; 10 Pairs/Cs
₹57,900.00INR / Case of 10
COMPARE
White Cleanroom Safety Shoe, White, Size 43; 10 Pairs/Cs
₹57,900.00INR / Case of 10
COMPARE
White Cleanroom Safety Shoe, White, Size 45; 10 Pairs/Cs
₹57,900.00INR / Case of 10
COMPARE
White Cleanroom Safety Shoe, White, Size 47; 10 Pairs/Cs
₹57,900.00INR / Case of 10
COMPARE
White Cleanroom Safety Shoe, White, Size 48; 10 Pairs/Cs
₹57,900.00INR / Case of 10
COMPARE
Black Cleanroom Safety Shoe, Black, Size 36, 10 Pairs/Case
₹61,000.00INR / Case of 10
COMPARE
Black Cleanroom Safety Shoe, Black, Size 37, 10 Pairs/Case
₹61,000.00INR / Case of 10
COMPARE
Black Cleanroom Safety Shoe, Black, Size 38, 10 Pairs/Case
₹61,000.00INR / Case of 10
COMPARE
Black Cleanroom Safety Shoe, Black, Size 39, 10 Pairs/Case
₹61,000.00INR / Case of 10
COMPARE
Black Cleanroom Safety Shoe, Black, Size 40, 10 Pairs/Case
₹61,000.00INR / Case of 10
COMPARE
Black Cleanroom Safety Shoe, Black, Size 41, 10 Pairs/Case
₹61,000.00INR / Case of 10
COMPARE
Black Cleanroom Safety Shoe, Black, Size 42, 10 Pairs/Case
₹61,000.00INR / Case of 10
Showing 1 - 20 of 26