Cole-Parmer® Cleanroom Paper
Autoclavable cleanroom paper for cleanroom environments where the prevention of chemical exportation and particle contamination is critical
Features:
• Autoclavable
• Specialized coating to reduce the release of particles, fibres and micro-organisms
• RoHS guidelines Compliant
• Weight: 80 GSM
• Colour: White, Blue
• Dimensions: A4 (297 x 210 mm)
• Retains ink and toner to reduce smudging
• Excellent heat resistant material for laser printing and photocopiers
• Designed for use in ISO Class 5 - 7 cleanrooms
• Lot to Lot traceable
8,300.00 - ₹10,500.00INR / Case of 2500
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Length (in) Width (in) Description
Availability Pricing
COMPARE
11.69 8.27 Cole-Parmer Cleanroom Paper, A4, 80 GSM, White; 250 sheets/Pk,10 Pk/Cs
₹8,300.00INR / Case of 2500
COMPARE
11.69 8.27 Cole-Parmer Cleanroom Paper, A4, 80 GSM, Blue; 250 sheets/Pk,10 Pk/Cs
₹10,500.00INR / Case of 2500