Cole-Parmer® Chain Clamps
Provide support for your specific laboratory equipment
Chain clamps have a quick slip-on-chain connection and a large easy-to-turn adjustment knob. Holds circular and irregularly shaped apparatus firmly. Clamps include integral connector and slip-on vinyl sleeve. NOTE: An additional connector must be purchased in order to attach clamps to frames or other apparatus.
33,300.00 - ₹84,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Clamp Material Length (in) Description
Availability Pricing
COMPARE
Nickel-plated Zinc with red vinyl and black fiberglass sleeves 7.4 Chain Clamp, Small, 6-11/16" (170 mm) Dia, Nickel-Plated Zinc
₹33,300.00INR / Each
COMPARE
Nickel-plated Zinc with red vinyl and black fiberglass sleeves 8.1 Chain Clamp, Large, 11" (280 mm) Dia, Nickel-Plated Zinc
₹50,500.00INR / Each
COMPARE
Nickel-plated Zinc with red vinyl and black fiberglass sleeves 15.1 Chain Clamp, 11" Dia, 12" Ext Arm; Nickel-Plated Zinc
₹35,700.00INR / Each
COMPARE
Stainless Steel 8.11 Chain Clamp, Large, 11" (280 mm) Dia, Stainless Steel
₹84,000.00INR / Each