Cole-Parmer Chain Clamp, Small, 6-11/16" (170 mm) Dia, Nickel-Plated Zinc
Representative image only.
Cole-Parmer Chain Clamp, Small, 6-11/16" (170 mm) Dia, Nickel-Plated Zinc
Cole-Parmer | Item: 0804732
Provide support for your specific laboratory equipment
₹33,300.00INR / Each
₹33,300.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Chain clamps have a quick slip-on-chain connection and a large easy-to-turn adjustment knob. Holds circular and irregularly shaped apparatus firmly. Clamps include integral connector and slip-on vinyl sleeve. NOTE: An additional connector must be purchased in order to attach clamps to frames or other apparatus.
1 Year Warranty
Specifications
  • Clamp MaterialNickel-plated Zinc with red vinyl and black fiberglass sleeves
  • Clamp TypeEquipment, Chain
  • Min Grip Range (InchShort)1.38
  • Max Grip Range (InchShort)6.6875
  • Length (in)7.4
  • DescriptionChain Clamp, Small, 6-11/16" (170 mm) Dia, Nickel-Plated Zinc