Cole-Parmer® CELLroll Cell Cultivation System Accessories
Customized configuration of CELLROLL increases flexibility and efficiency
  • Enhance performance of your CELLROLL system with modular additions
  • Optimize speed and bottle capacity with these CELLROLL components and accessories
  • CELLROLL system sold separately
The modular design of the CELLROLL cell cultivation system allows scale-up of bottle capacity from 4 to 16 roller bottles. Add additional storage decks to expend vertically or expand horizontally with horizontal storage decks. By using two drive units up to 32 roller bottles can be operated using one control unit only.
Successful growth of cells in roller bottles often relies on optimal rolling speed and therefore the CELLROLL allows fine tuning of the rotation speed. The standard drive unit operates in a speed range from 0.1 to 2 rpm and is adjustable in steps of 0.1 rpm. The faster drive unit rotates the roller bottles with 2 to 6 rpm (adjustable in steps of 0.5 rpm).
The customized configuration of CELLROLL increases flexibility and efficiency of the lab. The CELLROLL cell cultivation systems are sold separately as models 13050-00 and 13050-02.
256,000.00 - ₹654,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Number of Bottles Accepted Description
Availability Pricing
COMPARE
2 Additional Storage Deck for Vertical Extension
₹256,000.00INR / Each
COMPARE
16 Standard Drive Unit, 0.1 to 2.0 rpm
₹559,000.00INR / Each
COMPARE
16 Fast Drive Unit, 2.0 6.0 rpm
₹654,000.00INR / Each