Cole-Parmer® Cell and Tissue Culture Flasks
Optimize the cultivation of cells and tissues in your lab with Cole-Parmer flasks
  • Designed for consistency, cell health and reproducibility, with surface and ancillary options
  • Innovative angled neck design offers easy pipetting and cell scraper access
  • Select from non-treated and surface-treated flasks with growth areas ranging from 12.5 to 300cm2
  • Corrugation free and smooth flask surface maximizes usable growth area
  • Upper triangular and wider base shape provides stability
  • Protuding ridges on the back side of the flask makes it easy for stacking
  • Gamma sterilized with strict integrity testing
  • Certified DNase-, RNase-free, and non-pyrogenic
  • Graduation marked on both sides along with special area near neck for marking
  • 2 different cap styles can be used in both open and closed system
  • Plug sealing caps - Standard polyethylene caps providing liquid and gas shear seal
  • Vented caps - Vented polyethylene caps contain a 0.22μ hydrophobic filter to allow gas exchange and minimize risk of cross contamination
  • Packaged in durable zip re-sealable self-standing plastic bags that allow flasks to remain upright and reduce contamination
  • Inner packaging is individually labelled for lot traceability
10,900.00 - ₹21,300.00INR / Case of 1
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap Type Culture Area (cm²)
Availability Pricing
COMPARE
25 Standard
₹13,300.00INR / Case of 1
COMPARE
25 Vent
₹17,100.00INR / Case of 1
COMPARE
50 Standard
₹14,100.00INR / Case of 1
COMPARE
50 Vent
₹18,300.00INR / Case of 1
COMPARE
250 Standard
₹14,600.00INR / Case of 1
COMPARE
250 Vent
₹17,600.00INR / Case of 1
COMPARE
600 Standard
₹10,900.00INR / Case of 1
COMPARE
600 Vent
₹12,900.00INR / Case of 1
COMPARE
850 Standard
₹11,300.00INR / Case of 1
COMPARE
850 Vent
₹11,400.00INR / Case of 1
COMPARE
25 Standard 12.5
₹15,100.00INR / Case of 1
COMPARE
25 Vent 12.5
₹18,300.00INR / Case of 1
COMPARE
50 Standard 25
₹16,600.00INR / Case of 1
COMPARE
50 Vent 25
₹20,100.00INR / Case of 1
COMPARE
250 Standard 75
₹20,300.00INR / Case of 1
COMPARE
250 Vent 75
₹21,300.00INR / Case of 1
COMPARE
600 Standard 182
₹14,000.00INR / Case of 1
COMPARE
600 Vent 182
₹14,600.00INR / Case of 1
COMPARE
850 Standard 300
₹12,300.00INR / Case of 1
COMPARE
850 Vent 300
₹12,300.00INR / Case of 1
Showing 1- 20 of 20